148.81 GBP valid until 2025-12-31
Use promo code "24314" to get your promotional price.
Use promo code "24314" to get your promotional price.
As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.
| Accuracy | 0.6 to 1.8% |
|---|---|
| Compatible Tips | SureOne™ tips |
| Autoclavable | Autoclavable |
| For Use With | SureOne™ Tip |
| Product Line | Elite |
| Type | Micro Pipette |
| Volume (Metric) | 20 to 200 μL |
| Finger Style | Ergonomic |
| Includes | Calibration certificate, in-lab calibration tool |
| Increments | 0.2μL |
| Secure Tip Attachment | Soft touch tip ejection |
| Warranty | 3 Years |