Use promo code "25339" to get your promotional price.
Sodium borohydride, 98% min
Use promo code "25339" to get your promotional price.
| Quantity | 100 g |
|---|
Chemical Identifiers
| CAS | 16940-66-2 |
|---|---|
| Molecular Formula | BH4Na |
| Molecular Weight (g/mol) | 37.83 |
| MDL Number | MFCD00003518 |
| InChI Key | YOQDYZUWIQVZSF-UHFFFAOYSA-N |
| Synonym | Sodium tetrahydridoborate |
| IUPAC Name | sodium boranuide |
| SMILES | [BH4-].[Na+] |
Specification
| Chemical Name or Material | Sodium borohydride |
|---|---|
| Melting Point | 400°C (decomposition) |
| CAS | 16940-66-2 |
| Color | White |
| Physical Form | Granules |
| Quantity | 100 g |
| Assay Percent Range | 98% min. |
| Molecular Formula | BH4Na |
| MDL Number | MFCD00003518 |
| Synonym | Sodium tetrahydridoborate |
| Solubility Information | Soluble in water. Also soluble in liquid ammonia,amines,pyridine. |
| InChI Key | YOQDYZUWIQVZSF-UHFFFAOYSA-N |
| SMILES | [BH4-].[Na+] |
| IUPAC Name | sodium boranuide |
| Molecular Weight (g/mol) | 37.83 |
| Formula Weight | 37.83 |
| Ignition Point | 220°C (428°F) |
| Odor | Odorless |
| Sensitivity | Moisture sensitive |
| Density | 1.074 |
Technical documents
[Sodium borohydride]
[Danger]
[{code=H260, sequence=1, description=In contact with water releases flammable gases which may ignite spontaneously.}, {code=H301, sequence=2, description=Toxic if swallowed.}, {code=H314, sequence=3, description=Causes severe skin burns and eye damage.}, {code=H360, sequence=4, description=May damage fertility or the unborn child.}]
[{code=P201, sequence=51, description=Obtain special instructions before use.}, {code=P202, sequence=52, description=Do not handle until all safety precautions have been read and understood.}, {code=P223, sequence=53, description=Do not allow contact with water.}, {code=P231+P232, sequence=54, description=Handle under inert gas. Protect from moisture.}, {code=P260, sequence=55, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P264, sequence=56, description=Wash face, hands and anyexposed skin thoroughly after handling.}, {code=P270, sequence=57, description=Do not eat, drink or smoke when using this product.}, {code=P271, sequence=58, description=Use only outdoors or in a well-ventilated area.}, {code=P280, sequence=59, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P303+P361+P353, sequence=60, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=61, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=62, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=63, description=Immediately call a POISON CENTER/doctor/.}, {code=P330, sequence=64, description=Rinse mouth.}, {code=P331, sequence=65, description=Do NOT induce vomiting.}, {code=P363, sequence=66, description=Wash contaminated clothing before reuse.}, {code=P370+P378, sequence=67, description=In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.}, {code=P402+P404, sequence=68, description=Store in a dry place. Store in a closed container.}, {code=P405, sequence=69, description=Store locked up.}, {code=P501b, sequence=70, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: Sodium borohydride}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS06, sequence=3, description=SKULL & CROSSBONES}, {code=GHS08, sequence=4, description=HEALTH HAZARD}]
Product Identifier
- Sodium borohydride
Signal Word
- Danger
Hazard Statement
- H260- In contact with water releases flammable gases which may ignite spontaneously.
- H301- Toxic if swallowed.
- H314- Causes severe skin burns and eye damage.
- H360- May damage fertility or the unborn child.
Precautionary Statement
- P201- Obtain special instructions before use.
- P202- Do not handle until all safety precautions have been read and understood.
- P223- Do not allow contact with water.
- P231+P232- Handle under inert gas. Protect from moisture.
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P264- Wash face, hands and anyexposed skin thoroughly after handling.
- P270- Do not eat, drink or smoke when using this product.
- P271- Use only outdoors or in a well-ventilated area.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
- P330- Rinse mouth.
- P331- Do NOT induce vomiting.
- P363- Wash contaminated clothing before reuse.
- P370+P378- In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P402+P404- Store in a dry place. Store in a closed container.
- P405- Store locked up.
- P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental Information
- MIXTURE LIST- Contain: Sodium borohydride